missing translation for 'onlineSavingsMsg'
Learn More

Timosaponin AIII, MedChemExpress

MedChemExpress Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.

106.65 € - 384.75 €

Specifications

Chemical Name or Material Timosaponin AIII
CAS 41059-79-4
Color White
Purity Grade Notes Research
Molecular Formula C39H64O13
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16473906
Encompass_Preferred
View Documents
Medchem Express
HY-N0810
25 mg
384.75 €
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16441518
Encompass_Preferred
View Documents
Medchem Express
HY-N0810
5 mg
106.65 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16409118
Encompass_Preferred
View Documents
Medchem Express
HY-N0810
10 mg
186.30 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.
Specifications

Specifications

Timosaponin AIII
White
C39H64O13
C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@H](C)CO4)([H])C[C@@]1([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC2
740.92
Research
Cancer-programmed cell death
41059-79-4
Research
DMSO : 125 mg/mL (168.71 mM; Need ultrasonic)
740.92
98.0%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only