Learn More
Timosaponin BII, MedChemExpress
MedChemExpress Timosaponin BII (Prototimosaponin A III) is a steroid saponin found in the rhizomes of Anemarrhena asphodeloides. Timosaponin BII has neuronal protective, anti-inflammatory and antioxidant activities.
Brand:  Medchem Express HY-N0812
Encompass_Preferred
Product Code. 16451518
  • 112.05 € / 5mg
Explore more special offers
Description
Description
Timosaponin BII (Prototimosaponin A III) is a steroid saponin found in the rhizomes of Anemarrhena asphodeloides. Timosaponin BII has neuronal protective, anti-inflammatory and antioxidant activities.
Specifications
Specifications
Timosaponin BII
Off-White
C45H76O19
Prototimosaponin A III
C[C@@]1([C@]([C@@H]2C)([H])[C@](O[C@]2(O)CC[C@H](C)CO[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3CO)([H])C4)[C@]4([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC1
921.07
Research
Neuroscience-Neurodegeneration
136656-07-0
Research
5 mg
DMSO : 100 mg/mL (108.57 mM; Need ultrasonic) ∣H2O : 100 mg/mL (108.57 mM; Need ultrasonic)
921.07
98.52%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Timosaponin BII, MedChemExpress