Learn More
Tin, 30 mesh, Reagent ACS, Granular, Thermo Scientific Chemicals
50.80 € - 201.00 €
Chemical Identifiers
CAS | 7440-31-5 |
---|---|
Molecular Formula | Sn |
Molecular Weight (g/mol) | 118.71 |
MDL Number | MFCD00133862 |
InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
Synonym | powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german |
PubChem CID | 5352426 |
ChEBI | CHEBI:32990 |
IUPAC Name | tin |
SMILES | [Sn] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10762294
|
Thermo Scientific Acros
420991000 |
100 g | Glass bottle |
50.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10331551
|
Thermo Scientific Acros
420995000 |
500 g | Glass bottle |
201.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
5352426 | |
tin |
Sn | |
MFCD00133862 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
CHEBI:32990 | |
[Sn] |
Specifications
Gray | |
231.9°C | |
7.3000g/mL | |
Sn | |
01,1168; 02,414; 11,519; 12,486; 13,298; 17,333 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
[Sn] | |
118.71 | |
CHEBI:32990 | |
≥99.5% | |
Granular Powder |
7440-31-5 | |
2270.0°C | |
Sn | |
MFCD00133862 | |
15, 9603 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
118.69 | |
ACS Reagent | |
Tin |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 231-141-8
missing translation for 'rtecsNumber' : XP7320000
missing translation for 'tsca' : TSCA
RUO – Research Use Only