missing translation for 'onlineSavingsMsg'
Learn More

Tin, 99.5%, powder, -100 mesh, Thermo Scientific Chemicals

44.22 € - 138.69 €

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10308940
View Documents Promotion Details
Thermo Scientific Acros
317811000
100 g Plastic bottle
44.22 €
100g
Please to purchase this item. Need a web account? Register with us today!
10678634
View Documents Promotion Details
Thermo Scientific Acros
317815000
500 g Plastic bottle
138.69 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]

Specifications

Gray
231.9°C
7.3000g/mL
Sn
MFCD00133862
15, 9603
Solubility in water: insoluble. Other solubilities: soluble in acids and hot potassium hydroxide, solutions
[Sn]
118.71
CHEBI:32990
99.5%
Tin, -100 mesh
7440-31-5
2270.0°C
99.5%
Sn
01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
118.69
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Identifier
  • Tin, powder, -325 mesh

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only