Learn More
Tin, 99.8%, powder, -325 mesh, Thermo Scientific Chemicals
45.78 € - 142.71 €
Chemical Identifiers
CAS | 7440-31-5 |
---|---|
Molecular Formula | Sn |
Molecular Weight (g/mol) | 118.71 |
MDL Number | MFCD00133862 |
InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
Synonym | powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german |
PubChem CID | 5352426 |
ChEBI | CHEBI:32990 |
IUPAC Name | tin |
SMILES | [Sn] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10125330
|
Thermo Scientific Acros
317801000 |
100 g | Plastic bottle |
45.78 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371324
|
Thermo Scientific Acros
317805000 |
500 g | Plastic bottle |
142.71 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
5352426 | |
tin |
Sn | |
MFCD00133862 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
CHEBI:32990 | |
[Sn] |
Specifications
Gray | |
231.9°C | |
7.3000g/mL | |
Sn | |
MFCD00133862 | |
15, 9603 | |
Solubility in water: insoluble. Other solubilities: soluble in acids and hot potassium hydroxide, solutions | |
[Sn] | |
118.71 | |
CHEBI:32990 | |
99.8% | |
Tin, -325 mesh |
7440-31-5 | |
2270.0°C | |
99.8% | |
Sn | |
01,1168; 02,414; 11,519; 12,486; 13,298; 17,333 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
118.69 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- Tin, powder, -325 mesh
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.