Learn More
Tin arsenide, 99.999% (metals basis), Thermo Scientific™
46.43 € - 587.93 €
Chemical Identifiers
CAS | 39332-13-3 |
---|---|
Molecular Formula | C8H10N4O2S |
Molecular Weight (g/mol) | 226.254 |
MDL Number | MFCD00148646 |
InChI Key | OPKFIKAYNXEJQA-UHFFFAOYSA-N |
Synonym | 1,2-propanediol, 3-9h-purin-6-ylthio, 3-9h-purin-6-ylsulfanyl propane-1,2-diol, ncistruc1_000545, ncistruc2_000494, 1, 3-purin-6-ylthio, ax 3, 1, 3-1h-purin-6-ylthio, 6-2,3-dihydroxypropyl thiopurine, 3-9h-purin-6-ylthio-1,2-propanediol, 3-7h-purin-6-ylsulfanyl propane-1,2-diol |
PubChem CID | 3579846 |
IUPAC Name | 3-(7H-purin-6-ylsulfanyl)propane-1,2-diol |
SMILES | C1=NC2=C(N1)C(=NC=N2)SCC(CO)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343458
|
Thermo Scientific Alfa Aesar
036363.03 |
1 g |
46.43 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323458
|
Thermo Scientific Alfa Aesar
036363.06 |
5 g |
159.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333458
|
Thermo Scientific Alfa Aesar
036363.14 |
25 g |
587.93 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
39332-13-3 | |
226.254 | |
OPKFIKAYNXEJQA-UHFFFAOYSA-N | |
3579846 | |
C1=NC2=C(N1)C(=NC=N2)SCC(CO)O |
C8H10N4O2S | |
MFCD00148646 | |
1,2-propanediol, 3-9h-purin-6-ylthio, 3-9h-purin-6-ylsulfanyl propane-1,2-diol, ncistruc1_000545, ncistruc2_000494, 1, 3-purin-6-ylthio, ax 3, 1, 3-1h-purin-6-ylthio, 6-2,3-dihydroxypropyl thiopurine, 3-9h-purin-6-ylthio-1,2-propanediol, 3-7h-purin-6-ylsulfanyl propane-1,2-diol | |
3-(7H-purin-6-ylsulfanyl)propane-1,2-diol |
Specifications
39332-13-3 | |
C8H10N4O2S | |
UN1557 | |
Insoluble in water. Slightly soluble in tin telluride | |
C1=NC2=C(N1)C(=NC=N2)SCC(CO)O | |
226.254 | |
193.61 | |
(metals basis) |
Pieces | |
MFCD00148646 | |
1,2-propanediol, 3-9h-purin-6-ylthio, 3-9h-purin-6-ylsulfanyl propane-1,2-diol, ncistruc1_000545, ncistruc2_000494, 1, 3-purin-6-ylthio, ax 3, 1, 3-1h-purin-6-ylthio, 6-2,3-dihydroxypropyl thiopurine, 3-9h-purin-6-ylthio-1,2-propanediol, 3-7h-purin-6-ylsulfanyl propane-1,2-diol | |
OPKFIKAYNXEJQA-UHFFFAOYSA-N | |
3-(7H-purin-6-ylsulfanyl)propane-1,2-diol | |
3579846 | |
99.999% | |
Tin arsenide |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
H301+H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures