missing translation for 'onlineSavingsMsg'
Learn More

Tin Foil, 0.025mm (0.001 in.) thick, hard, 99.9% (Metals basis), Thermo Scientific Chemicals

Form: Approx. 4.2g/15 x 15cm; Temper: as rolled

191.96 € - 554.76 €

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336887
SDS View Documents
Thermo Scientific Alfa Aesar
010789.VA
15 x 15 cm
191.96 €
Each
Please to purchase this item. Need a web account? Register with us today!
11346887
SDS View Documents
Thermo Scientific Alfa Aesar
010789.VE
15 x 75 cm
554.76 €
Each
Please to purchase this item. Need a web account? Register with us today!
11356887
SDS View Documents
Thermo Scientific Alfa Aesar
010789.CL
15 x 300 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]

Specifications

Silver to Gray
Silver Gray foil
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
118.71
CHEBI:32990
Foil
7440-31-5
99.9% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
Odorless
Tin, Hard
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-141-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.