missing translation for 'onlineSavingsMsg'
Learn More
Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals Product Code.: 10125090

Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals

Product Code. 10125090
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10125090

Brand: Thermo Scientific Acros 222571000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Gray
Melting Point 231.9°C
Boiling Point 2270.0°C
Density 7.3000g/mL
Linear Formula Sn
Quantity 100 g
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Merck Index 15, 9603
Formula Weight 118.69
Packaging Glass bottle
Particle Size 0.1 to 0.8mm
Form Granular Powder
Chemical Name or Material Tin, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465082 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number222571000,222575000
Lot NumberA0447680 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number222571000,222575000
Lot NumberA0440729 Certificate TypeCertificate of Analysis Date01/04/2022 Catalog Number222571000,222575000
Lot NumberA0408346 Certificate TypeCertificate of Analysis Date05/05/2020 Catalog Number222571000,222575000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.