missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) 2-ethylhexanoate, tech., Thermo Scientific Chemicals

Product Code. 11323447
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, lubricants, addition agents and stabilizers for transformer oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301-10-0
Molecular Formula C16H30O4Sn
Molecular Weight (g/mol) 405.122
MDL Number MFCD00002676
InChI Key KSBAEPSJVUENNK-UHFFFAOYSA-L
Synonym stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
PubChem CID 9318
IUPAC Name 2-ethylhexanoate;tin(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]

Specifications

Color Colorless to Yellow
Density 1.251 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.494
Quantity 2 kg
Solubility Information Miscible with water.
Formula Weight 405.11
Grade Technical
Physical Form Liquid
Chemical Name or Material Tin(II) 2-ethylhexanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.