missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) acetate, 95%, Thermo Scientific Chemicals

Product Code. 11328027
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tin(II) acetate is used as a precursor in the production of tin anode, Sn-Cu bimetallic nanoparticles, high surface area tin oxide catalysts. It acts as Lewis acid catalysts for the esterification and transesterification reactions. It serves as an efficient catalytic system for carbon-hydrogen activation of methoxy group of anisole in presence of palladium(II) acetate. It plays an important role in stannous oral care compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-39-1
Molecular Formula C4H6O4Sn
Molecular Weight (g/mol) 236.798
MDL Number MFCD00054408
InChI Key PNOXNTGLSKTMQO-UHFFFAOYSA-L
Synonym tin ii acetate, stannous acetate, unii-1sku167w8p, tin 2+ diacetate, tin diacetate, tin acetate, tin di acetate, stannous diacetate, diacetoxytin, tech-95
PubChem CID 69488
IUPAC Name tin(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Sn+2]

Specifications

Melting Point 182°C to 183°C
Color White
Physical Form Lump/Powder
Quantity 5 g
Assay Percent Range 95%
Solubility Information Decomposes in water. Soluble in dilute HClSoluble in dilute hydrochloric acid,dimethyl sulfoxide and methanol.
Formula Weight 236.8
Sensitivity Hygroscopic
Appearance White powder
Density 2.31
Chemical Name or Material Tin(II) acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'