missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) bromide, 99.2%, Thermo Scientific Chemicals

Product Code. 11375727
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Stannous bromide acts a lewis acid. It is used to prepare alkyl tin compounds and in the spectrophotometric determination of iridium. It is involved in regioselective mono-etherfications of vicinal diols with diazo compounds. It acts as a catalyst to produce resinous lactone high polymers. It serves as a potent reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-24-0
Molecular Formula SnBr2
MDL Number MFCD00011239
Synonym tin ii bromide, tin dibromide, stannous bromide, tin bromide snbr2, snbr2, acmc-1bp22

Specifications

Boiling Point 623°C
Melting Point 215°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.4%
Solubility Information Soluble in alcohol,ether,acetone,pyridine and dimethyl sulfoxide.
Formula Weight 278.51
Density 5.12
Chemical Name or Material Tin(II) bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'