missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) bromide, 99.2%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Stannous bromide acts a lewis acid. It is used to prepare alkyl tin compounds and in the spectrophotometric determination of iridium. It is involved in regioselective mono-etherfications of vicinal diols with diazo compounds. It acts as a catalyst to produce resinous lactone high polymers. It serves as a potent reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 623°C |
Melting Point | 215°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.4% |
Solubility Information | Soluble in alcohol,ether,acetone,pyridine and dimethyl sulfoxide. |
Formula Weight | 278.51 |
Density | 5.12 |
Chemical Name or Material | Tin(II) bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'