missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) bromide, 99.2%

Product Code. p-4407992
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11375727

Brand: Thermo Scientific Alfa Aesar 043430.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-24-0 | SnBr2

Stannous bromide acts a lewis acid. It is used to prepare alkyl tin compounds and in the spectrophotometric determination of iridium. It is involved in regioselective mono-etherfications of vicinal diols with diazo compounds. It acts as a catalyst to produce resinous lactone high polymers. It serves as a potent reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-24-0
Molecular Formula SnBr2
MDL Number MFCD00011239
Synonym tin ii bromide, tin dibromide, stannous bromide, tin bromide snbr2, snbr2, acmc-1bp22

Specifications

Boiling Point 623°C
Melting Point 215°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.4%
Solubility Information Soluble in alcohol,ether,acetone,pyridine and dimethyl sulfoxide.
Formula Weight 278.51
Density 5.12
Chemical Name or Material Tin(II) bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.