missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Product Code. 10512991
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652.0°C
Melting Point 246.0°C
Color White
Physical Form Fine Crystalline Powder or Flakes
Quantity 25 g
Assay Percent Range 97.5 to 102.5% (Iodimetry)
Linear Formula SnCl2
Fieser 01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334
Merck Index 15, 8910
Solubility Information Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol
IUPAC Name λ²-tin(2+) dichloride
Formula Weight 189.6
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 3.95
Density 3.9500g/mL
Chemical Name or Material Tin(II) chloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'