missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals Product Code.: 10572791

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Product Code. 10572791
100 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10572791

Brand: Thermo Scientific Acros 196981000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652.0°C
Melting Point 246.0°C
Color White
Physical Form Fine Crystalline Powder or Flakes
Quantity 100 g
Assay Percent Range 97.5 to 102.5% (Iodimetry)
Linear Formula SnCl2
Fieser 01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334
Merck Index 15, 8910
Solubility Information Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol
IUPAC Name dichlorotin
Formula Weight 189.6
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 3.95
Density 3.9500g/mL
Chemical Name or Material Tin(II) chloride, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463327 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number196981000,196980250,196985000,196980025
Lot NumberA0442782 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number196981000,196980250,196985000,196980025
Lot NumberA0435084 Certificate TypeCertificate of Analysis Date26/05/2023 Catalog Number196981000,196980250,196985000,196980025
Lot NumberA0452005 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number196981000,196980250,196985000,196980025
Lot NumberA0394658 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number196981000,196980250,196985000,196980025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tin(II) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.