Learn More
Tin(II) chloride, anhydrous, 99% min, Thermo Scientific Chemicals
99.70 € - 1186.91 €
Chemical Identifiers
CAS | 7772-99-8 |
---|---|
Molecular Formula | Cl2Sn |
Molecular Weight (g/mol) | 189.61 |
MDL Number | MFCD00011241 |
InChI Key | AXZWODMDQAVCJE-UHFFFAOYSA-L |
Synonym | Stannous chloride |
PubChem CID | 24479 |
ChEBI | CHEBI:78067 |
SMILES | [Cl-].[Cl-].[Sn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314957
|
Thermo Scientific Alfa Aesar
041960.30 |
250 g |
99.70 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394947
|
Thermo Scientific Alfa Aesar
041960.A1 |
1 kg |
325.62 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304957
|
Thermo Scientific Alfa Aesar
041960.A7 |
5 kg |
1186.91 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It acts as an antisludging agent and a stabilizer. It can be used to reduce certain metal salts into metals, tin plating and in analytical chemistry to test mercury ions. It is also used as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. It is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7772-99-8 | |
189.61 | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
24479 | |
[Cl-].[Cl-].[Sn++] |
Cl2Sn | |
MFCD00011241 | |
Stannous chloride | |
CHEBI:78067 |
Specifications
652°C | |
7772-99-8 | |
99% min | |
MFCD00011241 | |
Soluble in water,alkalies,alcohol,methy lethyl ketone,methyl acetate,and acetone. | |
[Cl-].[Cl-].[Sn++] | |
189.61 | |
CHEBI:78067 | |
Air sensitive; Hygroscopic | |
Tin(II) chloride, anhydrous |
247°C | |
Crystalline | |
Cl2Sn | |
Stannous chloride | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
λ²-tin(2+) dichloride | |
24479 | |
189.62 | |
3.95 g/mL |
Safety and Handling
P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
H290-H302+H332-H314-H317-H335-H373
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 231-868-0
missing translation for 'rtecsNumber' : XP8700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Tin(II) chloride, anhydrous
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Stannous chloride
RUO – Research Use Only