missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) Chloride Dihydrate, 98%, ACS Reagent, Honeywell Fluka™
ACS Reagent, 98%
33.15 € - 2841.39 €
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15682620
|
Honeywell-Fluka
243523-5G |
5 g | Poly Bottle | ||||||
15642620
|
Honeywell-Fluka
243523-100G |
100 g | Poly Bottle | ||||||
15672620
|
Honeywell-Fluka
243523-500G |
500 g | Poly Bottle | ||||||
15662620
|
Honeywell-Fluka
243523-2.5KG |
2.5 kg | Poly Bottle |
859.95 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15652620
|
Honeywell-Fluka
243523-12KG |
12 kg | Drum with inliner |
2841.39 €
12kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10025-69-1 | |
MFCD00149863 |
Specifications
652°C | |
10025-69-1 | |
SnCl2 · 2H2O | |
UN3260 | |
225.65g/mol | |
ACS Reagent | |
2.71g/cm3 |
37°C to 38°C | |
Colorless | |
MFCD00149863 | |
Soluble in water, Hydrochloric Acid | |
98% | |
Crystalline | |
Tin(II) Chloride Dihydrate |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314-H317-H332-H335-H341-H361-H373-H410
missing translation for 'dotInformation' : 8/PGII
missing translation for 'einecsNumber' : 231-868-0
missing translation for 'rtecsNumber' : XP8850000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tin(II) chloride dihydrate
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.