missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10407850
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
MDL Number 149863

Specifications

Boiling Point 652°C
Melting Point 37°C
Color Colorless
Physical Form Solid
Quantity 500 g
Formula Weight 225.63g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'