missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride, ultra dry, 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 11378577
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It acts as an antisludging agent and a stabilizer. It can be used to reduce certain metal salts into metals, tin plating and in analytical chemistry to test mercury ions. It is also used as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. It is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Melting Point 247°C
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water,alkalies,alcohol,methyl ethyl ketone,methyl acetate and acetone.
IUPAC Name λ²-tin(2+) dichloride
Formula Weight 189.62
Sensitivity Air sensitive; Hygroscopic
Density 3.95
Chemical Name or Material Tin(II) Chloride, Ampouled under argon
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'