Learn More
Tin(II) ethoxide, Thermo Scientific Chemicals
46.13 € - 613.00 €
Chemical Identifiers
CAS | 14791-99-2 |
---|---|
Molecular Formula | C4H10O2Sn |
Molecular Weight (g/mol) | 208.832 |
MDL Number | MFCD00015653 |
InChI Key | XCKWFNSALCEAPW-UHFFFAOYSA-N |
Synonym | tin ii ethoxide, diethoxytin ii, acmc-20aji5, ethanol, tin 2+ salt 9ci |
PubChem CID | 71317418 |
IUPAC Name | ethanolate;tin(2+) |
SMILES | CC[O-].CC[O-].[Sn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410510
|
Thermo Scientific Alfa Aesar
020896.03 |
1 g |
46.13 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337977
|
Thermo Scientific Alfa Aesar
020896.06 |
5 g |
150.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347977
|
Thermo Scientific Alfa Aesar
020896.14 |
25 g | N/A | ||||||
Description
Tin(II) ethoxide is used in catalyst synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14791-99-2 | |
208.832 | |
XCKWFNSALCEAPW-UHFFFAOYSA-N | |
71317418 | |
CC[O-].CC[O-].[Sn+2] |
C4H10O2Sn | |
MFCD00015653 | |
tin ii ethoxide, diethoxytin ii, acmc-20aji5, ethanol, tin 2+ salt 9ci | |
ethanolate;tin(2+) |
Specifications
14791-99-2 | |
C4H10O2Sn | |
MFCD00015653 | |
tin ii ethoxide, diethoxytin ii, acmc-20aji5, ethanol, tin 2+ salt 9ci | |
XCKWFNSALCEAPW-UHFFFAOYSA-N | |
ethanolate;tin(2+) | |
71317418 | |
Solid/Semi-solid |
∼200°C (decomposition) | |
Sn(OC2H5)2 | |
Air and moisture sensitive | |
Reacts with water | |
CC[O-].CC[O-].[Sn+2] | |
208.832 | |
208.81 | |
Tin(II) ethoxide |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.