missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) fluoride, 97.5%

Product Code. p-7048814
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11337247

Brand: Thermo Scientific Alfa Aesar 011544.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-47-3

Tin(II) fluoride is used in the prevention of dental caries. It is an active ingredient in toothpastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-47-3
MDL Number MFCD00042540

Specifications

Boiling Point 850°C
Melting Point 219°C
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula SnF2
UN Number UN3260
Merck Index 14,8784
Solubility Information Soluble in potassium hydroxide and potassium fluoride. Insoluble in water,ethanol,ether and chloroform.
Formula Weight 156.69
Percent Purity 97.5%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 4.57 g/mL
Chemical Name or Material Tin(II) fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tin(II) fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Stannous fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.