missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) iodide, 99+%, Thermo Scientific Chemicals

Product Code. 11314809
Click to view available options
Quantity:
10 g
50 g

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3, which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-70-9
Molecular Formula I2Sn
Molecular Weight (g/mol) 372.519
MDL Number MFCD00049545
InChI Key JTDNNCYXCFHBGG-UHFFFAOYSA-L
Synonym tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 25138
IUPAC Name diiodotin
SMILES [Sn](I)I

Specifications

Boiling Point 720°C (decomposition)
Melting Point 320°C
Physical Form Powder
Quantity 10 g
Assay Percent Range >99%
Solubility Information Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides. Decomposes in water
Formula Weight 372.5
Sensitivity Air, light, and moisture sensitive
Density 5.28
Chemical Name or Material Tin(II) iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'