missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11302428

Tin(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11302428
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11302428

Brand: Thermo Scientific Alfa Aesar 014476.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3 which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-70-9
Molecular Formula I2Sn
Molecular Weight (g/mol) 372.519
MDL Number MFCD00049545
InChI Key JTDNNCYXCFHBGG-UHFFFAOYSA-L
Synonym tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 25138
IUPAC Name diiodotin
SMILES [Sn](I)I

Specifications

Boiling Point 720°C (decomposition)
Melting Point 320°C
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.99%, metal basis
Solubility Information Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides.
Formula Weight 372.5
Sensitivity Air and moisture sensitive
Density 5.28
Chemical Name or Material Tin(II) Iodide, Ampouled under argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.