Learn More
Tin(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
212.06 € - 739.68 €
Chemical Identifiers
CAS | 10294-70-9 |
---|---|
Molecular Formula | I2Sn |
Molecular Weight (g/mol) | 372.519 |
MDL Number | MFCD00049545 |
InChI Key | JTDNNCYXCFHBGG-UHFFFAOYSA-L |
Synonym | tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis |
PubChem CID | 25138 |
IUPAC Name | diiodotin |
SMILES | [Sn](I)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302428
|
Thermo Scientific Alfa Aesar
014476.06 |
5 g |
212.06 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392418
|
Thermo Scientific Alfa Aesar
014476.14 |
25 g |
739.68 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3 which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-70-9 | |
372.519 | |
JTDNNCYXCFHBGG-UHFFFAOYSA-L | |
25138 | |
[Sn](I)I |
I2Sn | |
MFCD00049545 | |
tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis | |
diiodotin |
Specifications
720°C (decomposition) | |
10294-70-9 | |
99.99%, metal basis | |
MFCD00049545 | |
Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides. | |
[Sn](I)I | |
372.519 | |
372.5 | |
5.28 |
320°C | |
Beads | |
I2Sn | |
tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis | |
JTDNNCYXCFHBGG-UHFFFAOYSA-L | |
diiodotin | |
25138 | |
Air and moisture sensitive | |
Tin(II) Iodide, Ampouled under argon |
Safety and Handling
When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 233-667-3
missing translation for 'rtecsNumber' : XQ3650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only