missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) methoxide, Thermo Scientific Chemicals

120.00 € - 432.00 €

Chemical Identifiers

CAS 14794-99-1
Molecular Formula C2H6O2Sn
Molecular Weight (g/mol) 180.778
MDL Number MFCD00015612
InChI Key UISUQHKSYTZXSF-UHFFFAOYSA-N
Synonym dimethoxytin ii, tin ii methoxide, acmc-1bxxn, methanol, tin 2+ salt 8ci,9ci
PubChem CID 71317297
IUPAC Name methanolate;tin(2+)
SMILES C[O-].C[O-].[Sn+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367977
View Documents
Thermo Scientific Alfa Aesar
020897.06
5 g
120.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11357977
View Documents
Thermo Scientific Alfa Aesar
020897.14
25 g
432.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14794-99-1
180.778
UISUQHKSYTZXSF-UHFFFAOYSA-N
71317297
C[O-].C[O-].[Sn+2]
C2H6O2Sn
MFCD00015612
dimethoxytin ii, tin ii methoxide, acmc-1bxxn, methanol, tin 2+ salt 8ci,9ci
methanolate;tin(2+)

Specifications

14794-99-1
C2H6O2Sn
Moisture sensitive
UISUQHKSYTZXSF-UHFFFAOYSA-N
methanolate;tin(2+)
71317297
Powder
242°C to 243°C
MFCD00015612
dimethoxytin ii, tin ii methoxide, acmc-1bxxn, methanol, tin 2+ salt 8ci,9ci
C[O-].C[O-].[Sn+2]
180.778
180.76
Tin(II) methoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.