missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxalate, 98%, Thermo Scientific Chemicals

Product Code. 11429193
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-94-8
Molecular Formula C2O4Sn
Molecular Weight (g/mol) 206.728
MDL Number MFCD00040678
InChI Key OQBLGYCUQGDOOR-UHFFFAOYSA-L
Synonym stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
PubChem CID 13149
IUPAC Name oxalate;tin(2+)
SMILES C(=O)(C(=O)[O-])[O-].[Sn+2]

Specifications

Melting Point 280°C (decomposition)
Quantity 100 g
Linear Formula SnC2O4
UN Number UN3288
Beilstein 3708588
Merck Index 14,8786
Formula Weight 206.72
Percent Purity 98%
Density 3.56
Chemical Name or Material Tin (II) oxalate
compliance-icons
Product Identifier
  • Stannous oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Stannous oxalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.