missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) sulfide, 99.5%, Thermo Scientific Chemicals

143.72 € - 493.46 €

Chemical Identifiers

CAS 1314-95-0
Molecular Formula SSn
Molecular Weight (g/mol) 150.77
MDL Number MFCD00011245
InChI Key DZXKSFDSPBRJPS-UHFFFAOYSA-N
Synonym tin ii sulfide, stannous sulfide, tin sulfide sns, tin monosulfide, tin sulphide, stannanethione, herzenbergite, zinnsulfur, acmc-1btyn
PubChem CID 426379
IUPAC Name λ²-tin(2+) sulfanediide
SMILES [S--].[Sn++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317667
SDS View Documents
Thermo Scientific Alfa Aesar
014051.06
5 g
143.72 €
5g
Please to purchase this item. Need a web account? Register with us today!
11327667
SDS View Documents
Thermo Scientific Alfa Aesar
014051.14
25 g
493.46 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Polymerization catalyst.Tin(II) sulfide is used as a polymerization catalyst. It is also used as a friction material. Further, it is used in optoelectronic device as well as a component in heterojunction photovoltaic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-95-0
150.77
DZXKSFDSPBRJPS-UHFFFAOYSA-N
426379
[S--].[Sn++]
SSn
MFCD00011245
tin ii sulfide, stannous sulfide, tin sulfide sns, tin monosulfide, tin sulphide, stannanethione, herzenbergite, zinnsulfur, acmc-1btyn
λ²-tin(2+) sulfanediide

Specifications

882°C
99.5%
MFCD00011245
Soluble in conc. HCl,hot conc. H2SO4.Insoluble in water.
[S--].[Sn++]
150.77
150.75
Tin(II) sulfide
1314-95-0
SSn
tin ii sulfide, stannous sulfide, tin sulfide sns, tin monosulfide, tin sulphide, stannanethione, herzenbergite, zinnsulfur, acmc-1btyn
DZXKSFDSPBRJPS-UHFFFAOYSA-N
λ²-tin(2+) sulfanediide
426379
5.08
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-248-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.