missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) tartrate hydrate, 95%, Thermo Scientific Chemicals

Product Code. 11304537
Click to view available options
Quantity:
25 g
100 g

It is used for dyeing and printing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815-85-0
Molecular Formula C4H4O6Sn
Molecular Weight (g/mol) 266.78
MDL Number MFCD00054358
InChI Key YXTDAZMTQFUZHK-UHFFFAOYNA-L
Synonym unii-6js9u5611x, stannous tartrate, tin tartrate, tartaric acid, tin 2+ salt 1:1, r-r*,r*-2,3-dihydroxybutanedioic acid, tin r-r*,r*-tartrate, butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1
PubChem CID 66545835
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioate;tin(2+)
SMILES [Sn++].OC(C(O)C([O-])=O)C([O-])=O

Specifications

Physical Form Powder
Quantity 25 g
Merck Index 14,8792
Solubility Information slightly soluble in water.
Formula Weight 266.76 (Anhydrous)
Percent Purity 95%
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Tin(II) tartrate hydrate
compliance-icons
Product Identifier
  • Tin(II) tartrate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Tin(II) tartrate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.