missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) tartrate hydrate, 95%, Thermo Scientific Chemicals

Product Code. 11314537
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used for dyeing and printing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815-85-0
Molecular Formula C4H4O6Sn
Molecular Weight (g/mol) 266.78
MDL Number MFCD00054358
InChI Key YXTDAZMTQFUZHK-UHFFFAOYNA-L
Synonym unii-6js9u5611x, stannous tartrate, tin tartrate, tartaric acid, tin 2+ salt 1:1, r-r*,r*-2,3-dihydroxybutanedioic acid, tin r-r*,r*-tartrate, butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1
PubChem CID 66545835
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioate;tin(2+)
SMILES [Sn++].OC(C(O)C([O-])=O)C([O-])=O

Specifications

Physical Form Powder
Quantity 100 g
Merck Index 14,8792
Solubility Information slightly soluble in water.
Formula Weight 266.76 (Anhydrous)
Percent Purity 95%
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Tin(II) tartrate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'