missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

117.59 € - 496.47 €

Chemical Identifiers

CAS 12040-02-7
Molecular Formula H2SnTe
Molecular Weight (g/mol) 248.33
MDL Number MFCD00049543
InChI Key UURRKPRQEQXTBB-UHFFFAOYSA-N
IUPAC Name stannanetellone
SMILES [SnH2]=[Te]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319409
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089186.09
10 g
117.59 €
10g
Please to purchase this item. Need a web account? Register with us today!
11309409
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089186.18
50 g
496.47 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) telluride is used as detector material for photodiodes in infrared used spectral range.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12040-02-7
248.33
UURRKPRQEQXTBB-UHFFFAOYSA-N
[SnH2]=[Te]
H2SnTe
MFCD00049543
stannanetellone

Specifications

780°C
Pieces
MFCD00049543
Insoluble in water.
[SnH2]=[Te]
248.33
99.999%
6.48 g/mL
Tin (II) telluride
12040-02-7
H2SnTe
UN3288
UURRKPRQEQXTBB-UHFFFAOYSA-N
stannanetellone
246.29
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 234-914-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only