missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) tetrafluoroborate, 50% w/w aq. soln.

Product Code. p-7080523
Click to view available options
Quantity:
1 kg
3 x 1 kg
Unit Size:
1kg
3kg
This item is not returnable. View return policy

Product Code. 11443810

Brand: Thermo Scientific Alfa Aesar 089669.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13814-97-6 | B2F8Sn | 292.32 g/mol

Tin(II) tetrafluoroborate is used in electroplating. It is also used for plating bath tinned copper, tin and tin alloy. Further, it acts as a catalyst for various organic reactions. It is also employed in metal plating. In addition to this, it is used as a petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13814-97-6
Molecular Formula B2F8Sn
Molecular Weight (g/mol) 292.32
MDL Number MFCD00054047
InChI Key JPZSNSDMGTYJIW-UHFFFAOYSA-N
Synonym tin ii fluoroborate, tin fluoroborate, tin 2+ ditetrafluoroborate, tin bis tetrafluoroborate, tin borofluoride, tin fluoborate, tin ii borofluoride, stannous tetrafluoroborate, tin ii tetrafluoroborate
PubChem CID 159698
IUPAC Name tin(2+);ditetrafluoroborate
SMILES [Sn++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 kg
Linear Formula Sn(BF4)2
UN Number UN3264
Solubility Information Miscible with water.
Formula Weight 292.3
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Density 1.670 g/mL
Chemical Name or Material Tin (II) tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.