missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Product Code. 11394527
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62086-04-8
Molecular Formula CH4F3O3SSn+
Molecular Weight (g/mol) 271.805
MDL Number MFCD00191251
InChI Key IACSYIHNPIQPMP-UHFFFAOYSA-M
Synonym tin ii trifluoromethanesulfonate
PubChem CID 131676109
IUPAC Name molecular hydrogen;tin(2+);trifluoromethanesulfonate
SMILES [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]

Specifications

Melting Point (decomposition)
Quantity 5 g
UN Number UN3288
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 416.82
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Tin(II) trifluoromethanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'