missing translation for 'onlineSavingsMsg'
Learn More

Tin ingot, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 15422107
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-31-5
Appearance Silver Gray ingot
Assay Percent Range 99.998% (Metals basis)
Molecular Formula Sn
MDL Number MFCD00133862
Quantity 1 kg
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES [Sn]
IUPAC Name tin
Molecular Weight (g/mol) 118.71
PubChem CID 5352426
ChEBI CHEBI:32990
Odor Odorless
Form Ingot
Chemical Name or Material Tin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'