missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) acetate, Thermo Scientific Chemicals

Product Code. 11473860
Click to view available options
Quantity:
5 g
25 g

Tin(IV) acetate is used to prepare tin-based oxides (Sn-Zn-P-O), which is a negative electrodes for rechargeable lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2800-96-6
Molecular Formula C8H12O8Sn
Molecular Weight (g/mol) 354.886
MDL Number MFCD00014976
InChI Key YJGJRYWNNHUESM-UHFFFAOYSA-J
Synonym tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
PubChem CID 9863446
IUPAC Name tin(4+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]

Specifications

Melting Point 232°C to 233°C
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in acetone and benzene.
Formula Weight 354.89
Sensitivity Hygroscopic
Chemical Name or Material Tin(IV) Acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'