missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

63.21 € - 228.14 €

Specifications

Chemical Name or Material Tin(IV) chloride
Name Note 1M Solution in Dichloromethane
CAS 7646-78-8, 75-09-2
Molecular Formula Cl4Sn
MDL Number MFCD00011242
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10629784
View Documents
Thermo Scientific Acros
378201000
100 mL AcroSeal™ Glass bottle
63.21 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10055992
View Documents
Thermo Scientific Acros
378208000
800 mL AcroSeal™ Glass Bottle
228.14 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Tin(IV) chloride
7646-78-8, 75-09-2
MFCD00011242
15, 8901
Solubility in water: reacts
Cl[Sn](Cl)(Cl)Cl
260.52
260.52
Clear colorless solution, fuming in air
1.4190g/mL
1M Solution in Dichloromethane
Cl4Sn
09,436; 17,335; 14,304; 13,300; 12,486; 10,370; 07,342; 06,553; 05,627; 03,269; 01,1111; 11,552
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
24287
1M, exact strength on the certificate of analysis
1.419
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Suspected of causing cancer.
May cause drowsiness or dizziness.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-588-9

Product Identifier
  • Tin(IV) chloride, 1M solution in dichloromethane

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only