missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) chloride, 99.999%, (trace metal basis)
Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
Packaging:
Glass bottle
Förpackningsstorlek
50g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acid
Spezifikation
Spezifikation
| Boiling Point | 114.0°C |
| Melting Point | -33.0°C |
| CAS | 7646-78-8 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 50 g |
| Assay Percent Range | 99.999% |
| Molecular Formula | Cl4Sn |
| Linear Formula | SnCl4 |
| MDL Number | MFCD00011242 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Tin(IV) chloride, anhydrous
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur