Learn More
Tin(IV) chloride, 99%, anhydrous, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 114.0°C |
Melting Point | -33.0°C |
Color | Brown-Red |
Physical Form | Solution |
Quantity | 100 mL |
Assay Percent Range | 98.5 to 101.5% (Argentometry) |
Linear Formula | SnCl4 |
Fieser | 01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 11,522; 12,486; 13,14; 14,304; 15,311; 17,335 |
Merck Index | 15,8901 |
Solubility Information | Solubility in water: reacts. Other solubilities: soluble in carbon disulfide, alcohol, carbon, tetrachloride, benzene, toluene, acetone, kerosene, and gasoline |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466182 | Certificate of Analysis | 19/09/2024 | 197510100,197510025,197510010,197511000 |
A0464755 | Certificate of Analysis | 26/07/2024 | 197510100,197510025,197510010,197511000 |
A0462976 | Certificate of Analysis | 23/05/2024 | 197510100,197510025,197510010,197511000 |
A0457802 | Certificate of Analysis | 14/11/2023 | 197510100,197510025,197510010,197511000 |
A0456203 | Certificate of Analysis | 08/09/2023 | 197510100,197510025,197510010,197511000 |
Safety and Handling


- Tin(IV) chloride, anhydrous
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.