missing translation for 'onlineSavingsMsg'
Learn More

Tin(Iv) Chloride Anhydrous, Pure, Fisher Chemical

Product Code. 10448420
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle

Specifications

Boiling Point 114°C
Melting Point -33°C
CAS 7646-78-8
Color Yellow
Physical Form Liquid
Quantity 100 mL
Molecular Formula Cl4Sn
MDL Number 11242
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
SMILES Cl[Sn](Cl)(Cl)Cl
IUPAC Name tetrachlorostannane
Molecular Weight (g/mol) 260.51
PubChem CID 24287
Formula Weight 260.52g/mol
Packaging Plastic Coated Glass bottle
Vapor Pressure 24.7mbar at 20°C
Density Vapor Density: 9
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tin(IV) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.