missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) fluoride, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11332158
Click to view available options
Quantity:
2 g
10 g

Friedel-Crafts catalyst

It is used as Friedel-Crafts catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-62-2
MDL Number MFCD00042541

Specifications

Melting Point 705°C (sublimation)
Physical Form Crystalline
Quantity 10 g
UN Number UN2923
Merck Index 14,8776
Formula Weight 194.68
Percent Purity 99%
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Tin(IV) fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.