missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) iodide, 95%, Thermo Scientific Chemicals

Product Code. 11324809
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in chemical research and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-47-8
Molecular Formula H4I4Sn
Molecular Weight (g/mol) 630.36
MDL Number MFCD00036272
InChI Key ZBCVLXKKLMNLGV-UHFFFAOYSA-N
Synonym stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
PubChem CID 24631
IUPAC Name tetraiodostannane
SMILES [Sn].I.I.I.I

Specifications

Boiling Point 340°C
Melting Point ∼143°C
Color Red
Physical Form Crystalline
Quantity 10 g
Linear Formula SnI4
UN Number UN3260
Merck Index 14,8777
Solubility Information Soluble in alcohol,benzene,CS2,chloroform,and ether; Decomposes in water
Formula Weight 626.31
Percent Purity 95%
Refractive Index 2.106
Sensitivity Moisture sensitive
Density 4.46 g/mL
Chemical Name or Material Tin (IV) iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'