missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene, Thermo Scientific Chemicals

41.41 € - 1290.42 €

Chemical Identifiers

CAS 1184-61-8
Molecular Formula C12H32O4Sn
Molecular Weight (g/mol) 359.094
MDL Number MFCD00145408
InChI Key VUDAJANAMZGVOP-UHFFFAOYSA-N
Synonym tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
PubChem CID 85354423
IUPAC Name propan-2-ol;tin
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377988
View Documents
Thermo Scientific Alfa Aesar
036563.AA
5 mL
41.41 €
5mL
Please to purchase this item. Need a web account? Register with us today!
11387988
View Documents
Thermo Scientific Alfa Aesar
036563.AC
25 mL
117.59 €
25mL
Please to purchase this item. Need a web account? Register with us today!
11367988
View Documents
Thermo Scientific Alfa Aesar
036563.AE
100 mL
351.75 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11475180
View Documents
Thermo Scientific Alfa Aesar
036563.AP
500 mL
1290.42 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1184-61-8
359.094
VUDAJANAMZGVOP-UHFFFAOYSA-N
85354423
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
C12H32O4Sn
MFCD00145408
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
propan-2-ol;tin

Specifications

1184-61-8
MFCD00145408
Moisture sensitive
Soluble in hydrocarbons,reacts with alcohols,ketones and esters
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
359.094
355.06
Liquid
Tin(IV) isopropoxide
C12H32O4Sn
UN1993
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
VUDAJANAMZGVOP-UHFFFAOYSA-N
propan-2-ol;tin
85354423
98%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H304-H314-H335-H336-H361d-H373

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.