missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∼130°C (decomposition) |
Boiling Point | 131°C (1.6 mmHg) |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in hydrocarbons and warm isopropanol. Hydrolyzes with water. Reacts with alcohols,ketones and esters. |
Formula Weight | 414.15 |
Percent Purity | 98% |
Physical Form | Wet Solid |
Assay | (metals basis) |
Chemical Name or Material | Tin(IV) isopropoxide isopropanol adduct |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'