missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, 15% in H{2}O colloidal dispersion, Thermo Scientific Chemicals

Product Code. 11495370
Click to view available options
Quantity:
500 g
2 kg

Tin(IV) oxide, O2Sn, CAS Number-18282-10-5, white tin oxide, tin iv oxide, tin peroxide, stannic dioxide, stannic oxide, tin oxide sno2, tin dioxide, stannic anhydride, cassiterite, flowers of tin, 500g, 15% in H2O colloidal dispersion, CHEBI:52991, 1.153 g/mL, 150.69, XOLBLPGZBRYERU-UHFFFAOYSA-N

It is used to detect carbon monoxide. Used in ceramic glazes and colors, putty, perfume preparations, textiles, polishing powder for steel, and the manufacture of special glasses. Stannic oxide is also used in making white patinas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name stannanedione
SMILES O=[Sn]=O

Specifications

Physical Form Liquid
Quantity 500 g
Assay Percent Range 15% in H2O colloidal dispersion
Solubility Information Fully miscible in water.
Formula Weight 150.69
Density 1.153 g/mL
Chemical Name or Material Tin(IV) oxide, pH 10.5-12.0, Viscosity 5-10cps, Particle charge ±negative. 1.15(Sp Grav)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'