missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

130.65 € - 392.96 €

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393378
SDS View Documents
Thermo Scientific Alfa Aesar
036302.14
25 g
130.65 €
25g
Please to purchase this item. Need a web account? Register with us today!
11383378
SDS View Documents
Thermo Scientific Alfa Aesar
036302.22
100 g
392.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For vacuum deposition. Used as a catalyst, in putty, as a polishing powder for steel and glass, in ceramic glazes and colors. The electrical properties of tin (iv) oxide render it an excellent material for the detection of very low levels of carbon monoxide, and several devices based on sintered pellets of SnO 2 are commercially available. Dye-sensitized fully solid state cells having the structure of rough n-type semiconductor film comprising zinc and tin (IV) oxide/Ru-bipyridyl complex/p-CuI are found to generate high short-circuit photocurrents and open-circuit voltages. The preparation of hydrous tin (IV) oxide in a suitable form for use as an ion exchanger is described. a colloidal-crystal templating technique to prepare three-dimensionally ordered macroporous (3DOM) tin (iv) oxide films for use as lithium ion anodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18282-10-5
150.71
XOLBLPGZBRYERU-UHFFFAOYSA-N
29011
dioxotin
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
CHEBI:52991
O=[Sn]=O

Specifications

1800°C to 1900°C (sublimation)
18282-10-5
Sintered Lump
SnO2
14,8778
Insoluble in water. Soluble in concentrated sulfuric acid,hydrochloric acid. Slowly soluble in hot concentrated potassium or sodium hydroxide solution.
O=[Sn]=O
150.71
CHEBI:52991
99.9%
1.9968
(metals basis)
1630°C
White
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
XOLBLPGZBRYERU-UHFFFAOYSA-N
dioxotin
29011
150.69
Odorless
6.95 g/mL
Tin(IV) oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 242-159-0

missing translation for 'rtecsNumber' : XQ4000000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.