missing translation for 'onlineSavingsMsg'
Learn More
Tin(IV) oxide, 99%, pure, Thermo Scientific Chemicals Product Code.: 15555126

Tin(IV) oxide, 99%, pure, Thermo Scientific Chemicals

Product Code. 15555126
1 kg
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
This item is not returnable. View return policy

Product Code. 15555126

Brand: Thermo Scientific Acros 451810010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydrideShow MoreShow Less
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O

Specifications

Quantity 1 kg
Linear Formula SnO2
Merck Index 15,8905
Solubility Information Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids
Formula Weight 150.69
Percent Purity 99%
Grade Pure
Specific Gravity 6.95
Density 6.9500g/mL
Chemical Name or Material Tin(IV) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456446 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number451810025,451810010,451811000
Lot NumberA0403781 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number451810025,451810010,451811000
Lot NumberA0410309 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number451810025,451810010,451811000
Lot NumberA0438758 Certificate TypeCertificate of Analysis Date24/02/2022 Catalog Number451810025,451810010,451811000
Lot NumberA0424018 Certificate TypeCertificate of Analysis Date02/11/2020 Catalog Number451810025,451810010,451811000
Product Identifier
  • Tin(IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(IV) oxide, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.