Learn More
Tin(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals
114.00 € - 405.00 €
Chemical Identifiers
CAS | 18282-10-5 |
---|---|
Molecular Formula | O2Sn |
Molecular Weight (g/mol) | 150.71 |
MDL Number | MFCD00011244 |
InChI Key | XOLBLPGZBRYERU-UHFFFAOYSA-N |
Synonym | tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride |
PubChem CID | 29011 |
ChEBI | CHEBI:52991 |
IUPAC Name | dioxotin |
SMILES | O=[Sn]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317067
|
Thermo Scientific Alfa Aesar
010891.06 |
5 g |
114.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307067
|
Thermo Scientific Alfa Aesar
010891.14 |
25 g |
405.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(IV) oxide is used as a pigment in the ceramic industry and as a polishing powder. It finds use in sensors of combustible gases. It is also used as a catalyst along with vanadium oxide for the oxidation of aromatic compounds to prepare carboxylic acids and acid anhydrides. It is used in sensors for detection of trace levels of carbon monoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18282-10-5 | |
150.71 | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
29011 | |
dioxotin |
O2Sn | |
MFCD00011244 | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
CHEBI:52991 | |
O=[Sn]=O |
Specifications
1800°C to 1900°C (sublimation) | |
18282-10-5 | |
99.996% (metals basis) | |
MFCD00011244 | |
Soluble in strong acids and alkali. Insoluble in water.Soluble in hot concentrated alkali and concentrated acids. Insoluble in water and alcohols. | |
O=[Sn]=O | |
150.71 | |
CHEBI:52991 | |
1.9968 | |
Tin(IV) oxide |
1630°C | |
Powder | |
O2Sn | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
dioxotin | |
29011 | |
150.69 | |
6.95 g/mL |
Safety and Handling
missing translation for 'einecsNumber' : 242-159-0
missing translation for 'rtecsNumber' : XQ4000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only