missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) tert-pentyloxide, 5% w/v in 2-methyl-2-butanol, 99% (metals basis), Thermo Scientific™

Product Code. 11347988
Click to view available options
Quantity:
5 mL
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00145409

Specifications

Boiling Point 100°C (0.2 mmHg)
Quantity 5 mL
UN Number UN3274
Sensitivity Moisture sensitive
Formula Weight 467.27
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tin(IV) tert-pentyloxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'