Learn More
Tin oxide, polymeric precursor, Oxide ≈29 wt%, Thermo Scientific™
Used in chemical sensors and transparent electrical conducting electrodes
361.00 €
Chemical Identifiers
CAS | 18282-10-5 |
---|---|
Molecular Formula | O2Sn |
Molecular Weight (g/mol) | 150.71 |
MDL Number | MFCD00011244 |
InChI Key | XOLBLPGZBRYERU-UHFFFAOYSA-N |
Synonym | tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride |
PubChem CID | 29011 |
ChEBI | CHEBI:52991 |
IUPAC Name | dioxotin |
SMILES | O=[Sn]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344727
|
Thermo Scientific Alfa Aesar
039753.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
18282-10-5 | |
150.71 | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
29011 | |
dioxotin |
O2Sn | |
MFCD00011244 | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
CHEBI:52991 | |
O=[Sn]=O |
Specifications
18282-10-5 | |
O2Sn | |
UN1993 | |
Soluble in alcohols | |
O=[Sn]=O | |
150.71 | |
CHEBI:52991 | |
1.59 g/mL |
Viscous Liquid | |
MFCD00011244 | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
dioxotin | |
29011 | |
Oxide ≈29 wt% | |
Tin oxide, Polymeric precursor |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures