Learn More
Tin plating powder, electroless, part A, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 044176.TD
130.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection
Specifications
Tin plating powder | |
MFCD03703330 | |
454 g |
Electroless, part A | |
UN1759 | |
Powder |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only