missing translation for 'onlineSavingsMsg'
Learn More

Tin powder, -100 mesh, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11311628
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Gray
Appearance Gray powder
Assay Percent Range 99.995%
Quantity 250 g
Mesh Size -100 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Tin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'